2,6-Dichloro(trifluoromethoxy)benzene

Lieferant: Apollo Scientific

Synonyme: 1,3-Dichloro-2-(trifluoromethoxy)benzene, 2,6-Dichloro-1-(trifluoromethoxy)benzene

PC408440-1G PC408440-5G
APOSPC408440-1GEA 288 EUR
APOSPC408440-1G APOSPC408440-5G
2,6-Dichloro(trifluoromethoxy)benzene
2,6-Dichloro(trifluoromethoxy)benzene
Formel: C₇H₃Cl₂F₃O
Molecular Weight: 231 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14697984
CAS-Nummer: 97608-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR