6-Chlor-2-fluorpurin 98%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-2-fluoro-9H-purine

PC408500-10G PC408500-100G PC408500-25G
APOSPC408500-10GEA 71.3 EUR
APOSPC408500-10G APOSPC408500-100G APOSPC408500-25G
6-Chlor-2-fluorpurin 98%
6-Chloro-2-fluoropurine
Formel: C₅H₂ClFN₄
Molecular Weight: 172,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02183557
CAS-Nummer: 1651-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR