tert-Butyl-4-(aminomethyl)-4-fluoropiperidine-1-carboxylate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
tert-Butyl 4-(aminomethyl)-4-fluoropiperidine-1-carboxylate, 1-Boc-4-(aminomethyl)-4-fluoropiperidine
PC408511-10G
PC408511-5G
APOSPC408511-10GEA
88.6
EUR
APOSPC408511-10G
APOSPC408511-5G
tert-Butyl-4-(aminomethyl)-4-fluoropiperidine-1-carboxylate
tert-Butyl-4-(aminomethyl)-4-fluoropiperidine-1-carboxylate
Formel:
C₁₁H₂₁FN₂O₂ Molecular Weight: 232.3 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11847970 CAS-Nummer: 620611-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...