1-(4-Fluorbenzyl)homopiperazin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC4089-1G
PC4089-10G
PC4089-5G
APOSPC4089-1GEA
29
EUR
APOSPC4089-1G
APOSPC4089-10G
APOSPC4089-5G
1-(4-Fluorbenzyl)homopiperazin 95%
1-(4-Fluorbenzyl)homopiperazin
Formel:
C₁₂H₁₇FN₂ Molecular Weight: 208,28 g/mol Siedepunkt: 112 °C/0,5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00816588 CAS-Nummer: 76141-89-4 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...