(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride
Lieferant: Apollo Scientific
Synonyme:
H-cis-4-F-Pro-NH₂·HCl, cis-4-Fluoro-L-prolinamide hydrochloride
PC409008-1G
PC409008-250MG
PC409008-5G
APOSPC409008-1GEA
422
EUR
APOSPC409008-1G
APOSPC409008-250MG
APOSPC409008-5G
(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride
(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride
Formel:
C₅H₁₀ClFN₂O Molecular Weight: 168.6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368567 CAS-Nummer: 426844-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...