(2-Chlor-4-fluorphenyl)essigsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Chloro-4-fluorophenyl)acetic acid

PC4091-100G PC4091-25G PC4091-5G
APOSPC4091-100GEA 156 EUR
APOSPC4091-100G APOSPC4091-25G APOSPC4091-5G
(2-Chlor-4-fluorphenyl)essigsäure 97%
(2-Chlor-4-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Molecular Weight: 188,59 g/mol
Schmelzpunkt: 102…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236028
CAS-Nummer: 177985-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR