2-Chlor-4-fluorphenylacetonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(2-Chloro-4-fluorophenyl)acetonitrile
PC4093-5G
PC4093-100G
PC4093-1G
APOSPC4093-5GEA
24.8
EUR
APOSPC4093-5G
APOSPC4093-100G
APOSPC4093-1G
2-Chlor-4-fluorphenylacetonitril 97%
2-Chlor-4-fluorphenylacetonitril
Formel:
C₈H₅ClFN Molecular Weight: 169,59 g/mol Schmelzpunkt: 67…69 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236029 CAS-Nummer: 75279-56-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...