4-Fluoronaphthalen-1-ol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC409501-1G
PC409501-10G
PC409501-5G
APOSPC409501-1GEA
426
EUR
APOSPC409501-1G
APOSPC409501-10G
APOSPC409501-5G
4-Fluoronaphthalen-1-ol 95+%
4-Fluoronaphthalen-1-ol
Formel:
C₁₀H₇FO Molecular Weight: 162.17 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699695 CAS-Nummer: 315-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...