3-(Trifluoromethyl)pyrrolidine-3-carboxylic acid 95+%
Lieferant: Apollo Scientific
PC409517-1G
PC409517-5G
APOSPC409517-1GEA
891
EUR
APOSPC409517-1G
APOSPC409517-5G
3-(Trifluoromethyl)pyrrolidine-3-carboxylic acid 95+%
3-(Trifluoromethyl)pyrrolidine-3-carboxylic acid
Formel:
C₆H₈F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258799 CAS-Nummer: 916423-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...