5-Hydrazinyl-2-(trifluoromethyl)pyridine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC409521-1G
PC409521-250MG
APOSPC409521-1GEA
132
EUR
APOSPC409521-1G
APOSPC409521-250MG
5-Hydrazinyl-2-(trifluoromethyl)pyridine 95+%
5-Hydrazinyl-2-(trifluoromethyl)pyridine
Formel:
C₆H₆F₃N₃ Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11113388 CAS-Nummer: 1035173-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...