6-(Trifluoromethyl)pyridin-3-yltrifluoromethanesulphonate 95+%
Lieferant: Apollo Scientific
PC409524-1G
PC409524-10G
PC409524-5G
APOSPC409524-1GEA
380
EUR
APOSPC409524-1G
APOSPC409524-10G
APOSPC409524-5G
6-(Trifluoromethyl)pyridin-3-yltrifluoromethanesulphonate 95+%
6-(Trifluoromethyl)pyridin-3-yltrifluoromethanesulphonate
Formel:
C₇H₃F₆NO₃S Molecular Weight: 295,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226775 CAS-Nummer: 1097778-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...