5-Isothiocyanato-2-(trifluoromethyl)pyridine 95+%

Lieferant: Apollo Scientific

PC409526-10G PC409526-1G PC409526-5G
APOSPC409526-10GEA 1030 EUR
APOSPC409526-10G APOSPC409526-1G APOSPC409526-5G
5-Isothiocyanato-2-(trifluoromethyl)pyridine 95+%
5-Isothiocyanato-2-(trifluoromethyl)pyridine
Formel: C₇H₃F₃N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD12401614
CAS-Nummer: 1208672-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR