(3-Fluor-4-(trifluormethyl)phenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-4-(trifluoromethyl)phenylacetic acid
PC4098-1G
PC4098-25G
PC4098-5G
APOSPC4098-1GEA
21.9
EUR
APOSPC4098-1G
APOSPC4098-25G
APOSPC4098-5G
(3-Fluor-4-(trifluormethyl)phenyl)essigsäure 98%
(3-Fluor-4-(trifluormethyl)phenyl)essigsäure
Formel:
C₉H₆F₄O₂ Molecular Weight: 222,14 g/mol Schmelzpunkt: 88…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236303 CAS-Nummer: 238754-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...