2,4-Diamino-5-fluorchinazolin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2,4-diaminoquinazoline

PC4099-5G
APOSPC4099-5GEA 157 EUR
APOSPC4099-5G
2,4-Diamino-5-fluorchinazolin
2,4-Diamino-5-fluoroquinazoline
Formel: C₈H₇FN₄
Molecular Weight: 178,17 g/mol
Schmelzpunkt: 245…248 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00221785
CAS-Nummer: 119584-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR