(4-Fluorphenoxy)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluorophenoxyacetic Acid

PC4100-100G PC4100-1G
APOSPC4100-100GEA 161 EUR
APOSPC4100-100G APOSPC4100-1G
(4-Fluorphenoxy)essigsäure
(4-Fluorphenoxy)essigsäure
Formel: (C₆H₄F)OCH₂COOH
Molecular Weight: 170.14 g/mol
Schmelzpunkt: 104…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004304
CAS-Nummer: 405-79-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR