2-(Brommethyl)-4-fluorbenzonitril
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromomethyl-4-fluorobenzonitrile
PC4108-25G
PC4108-5G
PC4108-100G
APOSPC4108-25GEA
82.8
EUR
APOSPC4108-25G
APOSPC4108-5G
APOSPC4108-100G
2-(Brommethyl)-4-fluorbenzonitril
2-Cyano-5-fluorobenzyl bromide
Formel:
C₈H₅BrFN Molecular Weight: 214,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059542 CAS-Nummer: 421552-12-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...