4-Bromo-1-(difluoromethyl)-5-methyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

PC411326-1G PC411326-25G PC411326-5G
APOSPC411326-1GEA 261 EUR
APOSPC411326-1G APOSPC411326-25G APOSPC411326-5G
4-Bromo-1-(difluoromethyl)-5-methyl-1H-pyrazole
4-Bromo-1-(difluoromethyl)-5-methyl-1H-pyrazole
Formel: C₅H₅BrF₂N₂
Molecular Weight: 211.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17078896
CAS-Nummer: 1243250-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR