(2-Fluoro-4-isopropoxyphenyl)boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-4-isopropoxyphenylboronic acid

PC412023-25G PC412023-5G PC412023-10G
APOSPC412023-25GEA 823 EUR
APOSPC412023-25G APOSPC412023-5G APOSPC412023-10G
(2-Fluoro-4-isopropoxyphenyl)boronic acid 98%
(2-Fluoro-4-isopropoxyphenyl)boronic acid
Formel: C₉H₁₂BFO₃
Molecular Weight: 197,9992 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17011998
CAS-Nummer: 586389-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR