5-Chlor-2-fluor-4-pyridinboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (5-Chloro-2-fluoropyridin-4-yl)boronic acid

PC412024-1G PC412024-5G
APOSPC412024-1GEA 59.6 EUR
APOSPC412024-1G APOSPC412024-5G
5-Chlor-2-fluor-4-pyridinboronsäure 97%
5-Chlor-2-fluor-4-pyridinboronsäure
Formel: C₅H₄BClFNO₂
Molecular Weight: 175.35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05662405
CAS-Nummer: 1034659-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR