4-Benzyloxy-2,6-difluorphenylboronsäure 95+%
Lieferant: Apollo Scientific
Synonyme:
(4-(Benzyloxy)-2,6-difluorophenyl)boronic acid
PC412025-5G
PC412025-10G
PC412025-1G
APOSPC412025-5GEA
220
EUR
APOSPC412025-5G
APOSPC412025-10G
APOSPC412025-1G
4-Benzyloxy-2,6-difluorphenylboronsäure 95+%
4-Benzyloxy-2,6-difluorphenylboronsäure
Formel:
C₁₃H₁₁BF₂O₃ Molecular Weight: 264,0325 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026734 CAS-Nummer: 156635-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...