5-Fluor-2-methoxy-4-pyridinboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-methoxypyridine-4-boronic acid

PC412027-5G
APOSPC412027-5GEA 122 EUR
APOSPC412027-5G
5-Fluor-2-methoxy-4-pyridinboronsäure 95%
5-Fluoro-2-methoxy-4-pyridineboronic acid
Formel: C₆H₇BFNO₃
Molecular Weight: 170,9300 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD06657887
CAS-Nummer: 1043869-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR