2-Brom-4-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromo-4-fluorophenyl)boronic acid

PC412058-1G PC412058-500MG PC412058-5G
APOSPC412058-1GEA 37.7 EUR
APOSPC412058-1G APOSPC412058-500MG APOSPC412058-5G
2-Brom-4-fluorphenylboronsäure 97%
2-Brom-4-fluorphenylboronsäure
Formel: C₆H₅BBrFO₂
Molecular Weight: 218,82 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13195647
CAS-Nummer: 1217501-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR