3-Cyan-4-fluorphenylboronsäurepinakolester 95+%
Lieferant: Apollo Scientific
Synonyme:
3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 2-(3-Cyano-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Cyano-4-fluorophenylboronic acid Pinacol ester, 3-Cyano-4-fluorophenylboronic acid, pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
PC412081-25G
APOSPC412081-25GEA
186
EUR
APOSPC412081-25G
3-Cyan-4-fluorphenylboronsäurepinakolester 95+%
3-Cyan-4-fluorphenylboronsäurepinakolester
Formel:
C₁₃H₁₅BFNO₂ Molecular Weight: 247,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06795681 CAS-Nummer: 775351-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...