5-Amino-2-fluorobenzeneboronic acid pinacol ester 95+%
Lieferant: Apollo Scientific
Synonyme:
4-FLUORO-3-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)ANILINE, 5-AMINO-2-FLUOROPHENYLBORONIC ACID PINACOL ESTER, 5-Amino-2-fluorophenylboronic acid pinacol ester
PC412222-5G
PC412222-1G
APOSPC412222-5GEA
699
EUR
APOSPC412222-5G
APOSPC412222-1G
5-Amino-2-fluorobenzeneboronic acid pinacol ester 95+%
5-Amino-2-fluorobenzeneboronic acid pinacol ester
Formel:
C₁₂H₁₇BFNO₂ Molecular Weight: 237,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16996293 CAS-Nummer: 1152441-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...