3-[3-(Trifluoromethyl)phenyl]-1H-pyrazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4123-1G
PC4123-250MG
APOSPC4123-1GEA
145
EUR
APOSPC4123-1G
APOSPC4123-250MG
3-[3-(Trifluoromethyl)phenyl]-1H-pyrazole 97%
3-[3-(Trifluoromethyl)phenyl]-1H-pyrazole
Formel:
C₁₀H₇F₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00973825 CAS-Nummer: 149739-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...