5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäure 96%
Lieferant: Apollo Scientific
PC412321-100MG
PC412321-250MG
APOSPC412321-100MGEA
725
EUR
APOSPC412321-100MG
APOSPC412321-250MG
5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäure 96%
5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäure
Formel:
C₈H₇BClF₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11053801 CAS-Nummer: 1072946-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...