8-(Trifluormethoxy)chinolin-5-boronsäure

Lieferant: Apollo Scientific

Synonyme: (8-(Trifluoromethoxy)quinolin-5-yl)boronic acid

PC412551-1G PC412551-250MG
APOSPC412551-1GEA 304 EUR
APOSPC412551-1G APOSPC412551-250MG
8-(Trifluormethoxy)chinolin-5-boronsäure
8-(Trifluormethoxy)chinolin-5-boronsäure
Formel: C₁₀H₇BF₃NO₃
Molecular Weight: 256.97 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11504851
CAS-Nummer: 1072951-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR