5-Fluor-2-iodanilin

Lieferant: Apollo Scientific
Achtung

PC4136-25G
APOSPC4136-25GEA 119 EUR
APOSPC4136-25G
5-Fluor-2-iodanilin
5-Fluor-2-iodanilin
Formel: C₆H₅FIN
Molecular Weight: 237,02 g/mol
Schmelzpunkt: 40…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093953
CAS-Nummer: 255724-71-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR