3-Fluorphenylhydraziniumchlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Fluorphenyl)hydrazin Hydrochlorid, 3-Fluorphenylhydrazin Hydrochlorid, 3-Fluorphenylhydrazinhydrochlorid

PC4154-100G PC4154-25G PC4154-500G
APOSPC4154-100GEA 87 EUR
APOSPC4154-100G APOSPC4154-25G APOSPC4154-500G
3-Fluorphenylhydraziniumchlorid 97%
3-Fluorphenylhydraziniumchlorid
Formel: C₆H₈ClFN₂
Molecular Weight: 162.59 g/mol
Schmelzpunkt: 268 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012934
CAS-Nummer: 2924-16-5
EINECS: 220-886-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR