4-Fluorphenylhydraziniumchlorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluorphenylhydrazin Hydrochlorid, 1-(4-Fluorphenyl)hydrazin Hydrochlorid, 4-Fluorphenylhydrazinhydrochlorid
PC4156-100G
PC4156-25G
PC4156-500G
APOSPC4156-100GEA
34.8
EUR
APOSPC4156-100G
APOSPC4156-25G
APOSPC4156-500G
4-Fluorphenylhydraziniumchlorid 97%
4-Fluorphenylhydraziniumchlorid
Formel:
C₆H₈ClFN₂ Molecular Weight: 162.59 g/mol Schmelzpunkt: 265 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00012942 CAS-Nummer: 823-85-8 EINECS: 212-521-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...