2-Fluor-α-methylbenzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC4171-5G PC4171-100G PC4171-25G
APOSPC4171-5GEA 33.5 EUR
APOSPC4171-5G APOSPC4171-100G APOSPC4171-25G
2-Fluor-α-methylbenzylalkohol 98%
2-Fluor-α-methylbenzylalkohol
Formel: C₈H₉FO
Molecular Weight: 140,16 g/mol
Siedepunkt: 110 °C/135mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00014401
CAS-Nummer: 445-26-1
EINECS: 207-155-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR