3-(4-Fluorophenyl)-5-(methylthio)pyrazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4174B-1G
PC4174B-250MG
PC4174B-10G
APOSPC4174B-1GEA
132
EUR
APOSPC4174B-1G
APOSPC4174B-250MG
APOSPC4174B-10G
3-(4-Fluorophenyl)-5-(methylthio)pyrazole 97%
3-(4-Fluorophenyl)-5-(methylthio)pyrazole
Formel:
C₁₀H₉FN₂S Molecular Weight: 208,26 g/mol Schmelzpunkt: 97…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052075 CAS-Nummer: 175137-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...