(4-Fluorphenylthio)essigsäure 97%
Lieferant: Apollo Scientific
Synonyme:
2-((4-Fluorophenyl)thio)acetic acid
PC4175D-10G
PC4175D-50G
APOSPC4175D-10GEA
119
EUR
APOSPC4175D-10G
APOSPC4175D-50G
(4-Fluorphenylthio)essigsäure 97%
(4-Fluorphenylthio)essigsäure
Formel:
C₈H₇FO₂S Molecular Weight: 186,21 g/mol Schmelzpunkt: 76…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00219825 CAS-Nummer: 332-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...