(4-Fluorphenylthio)aceton 97%

Lieferant: Apollo Scientific

Synonyme: (4-Fluorophenylthio)propan-2-one

PC4175H-1G PC4175H-5G
APOSPC4175H-1GEA 39.4 EUR
APOSPC4175H-1G APOSPC4175H-5G
(4-Fluorphenylthio)aceton 97%
(4-Fluorphenylthio)aceton
Formel: C₉H₉FOS
Molecular Weight: 184,23 g/mol
Siedepunkt: 100…103 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00179340
CAS-Nummer: 2968-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR