1-(4-Fluorphenyl)-2-thioharnstoff 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Fluorphenylthioharnstoff, N-(4-Fluorphenyl)thioharnstoff
PC4175P-5G
PC4175P-25G
PC4175P-100G
APOSPC4175P-5GEA
21.9
EUR
APOSPC4175P-5G
APOSPC4175P-25G
APOSPC4175P-100G
1-(4-Fluorphenyl)-2-thioharnstoff 98%
1-(4-Fluorphenyl)-2-thioharnstoff
Formel:
(C₆H₄F)NHC(S)NH₂ Molecular Weight: 170.21 g/mol Siedepunkt: 276 Schmelzpunkt: 162…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041180 CAS-Nummer: 459-05-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...