3-Fluorphthalsäureanhydrid 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Fluor-1,2-benzoldicarbonsäureanhydrid

PC4179N-100G PC4179N-25G PC4179N-5G
APOSPC4179N-100GEA 539 EUR
APOSPC4179N-100G APOSPC4179N-25G APOSPC4179N-5G
3-Fluorphthalsäureanhydrid 97%
3-Fluorphthalsäureanhydrid

Useful for the synthesis & bioactivity of photoaffinity labelof the plant growth regulator 1-(3-chlorophthalimido)cyclo-hexanecarboxamide: J.Agric.Food Chem., 39,554, (1991)

Formel: C₈H₃FO₃
Siedepunkt: 283 °C
Schmelzpunkt: 158…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039696
CAS-Nummer: 652-39-1
EINECS: 211-491-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR