Methyl-4-fluorsalicylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-fluoro-2-hydroxybenzoate

PC4187-25G
APOSPC4187-25GEA 183 EUR
APOSPC4187-25G
Methyl-4-fluorsalicylat 98%
Methyl-4-fluorsalicylat
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
Schmelzpunkt: 38…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093965
CAS-Nummer: 392-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR