(3,3-Difluorocyclobutyl)methanamine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC420001-1G PC420001-250MG
APOSPC420001-1GEA 626 EUR
APOSPC420001-1G APOSPC420001-250MG
(3,3-Difluorocyclobutyl)methanamine hydrochloride 97%
(3,3-Difluorocyclobutyl)methanamine hydrochloride
Formel: C₅H₁₀ClF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD12400850
CAS-Nummer: 1159813-93-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR