N,N-Dimethyl-2,2,2-trifluoracetamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2,2-Trifluoro-N,N-dimethylacetamide
PC4203-100G
PC4203-25G
APOSPC4203-100GEA
208
EUR
APOSPC4203-100G
APOSPC4203-25G
N,N-Dimethyl-2,2,2-trifluoracetamid
N,N-Dimethyl-2,2,2-trifluoracetamid
Formel:
C₄H₆F₃NO Molecular Weight: 141,0900 g/mol Siedepunkt: 134…136 °C Dichte: 1,264 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043555 CAS-Nummer: 1547-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...