3'-Chlor-5'-fluoracetophenon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Chloro-5-fluorophenyl)ethanone

PC4214-5G
APOSPC4214-5GEA 84.1 EUR
APOSPC4214-5G
3'-Chlor-5'-fluoracetophenon 97%
3'-Chlor-5'-fluoracetophenon
Formel: C₈H₆ClFO
Molecular Weight: 172,59 g/mol
Schmelzpunkt: 51…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115834
CAS-Nummer: 842140-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR