4-Chlor-3'-fluorbenzhydrol 97%

Lieferant: Apollo Scientific

PC4226-1G PC4226-5G
APOSPC4226-1GEA 161 EUR
APOSPC4226-1G APOSPC4226-5G
4-Chlor-3'-fluorbenzhydrol 97%
4-Chlor-3'-fluorbenzhydrol
Formel: C₁₃H₁₀ClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD06201293
CAS-Nummer: 2795-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR