4-(Trifluormethoxy)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Dihydroxy(4-(trifluormethoxy)phenyl)boran, 4-(Trifluormethoxy)benzolboronsäure, (4-(Trifluormethoxy)phenyl)metaborsäure

PC4241-25G PC4241-100G PC4241-5G
APOSPC4241-25GEA 26.1 EUR
APOSPC4241-25G APOSPC4241-100G APOSPC4241-5G
4-(Trifluormethoxy)phenylboronsäure 98%
4-(Trifluormethoxy)phenylboronsäure
Formel: C₇H₆BF₃O₃
Molecular Weight: 205.93 g/mol
Schmelzpunkt: 120…127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01074648
CAS-Nummer: 139301-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR