6-Fluorchromon-3-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC4261-1G
PC4261-5G
APOSPC4261-1GEA
29
EUR
APOSPC4261-1G
APOSPC4261-5G
6-Fluorchromon-3-carbonsäure
6-Fluorchromon-3-carbonsäure
Formel:
C₁₀H₅FO₄ Schmelzpunkt: 234…238 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424505 CAS-Nummer: 71346-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...