2-Chlor-6-fluorbenzotrifluorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chloro-3-fluoro-2-(trifluoromethyl)benzene
PC4269-1G
PC4269-5G
APOSPC4269-1GEA
127
EUR
APOSPC4269-1G
APOSPC4269-5G
2-Chlor-6-fluorbenzotrifluorid 98%
2-Chlor-6-fluorbenzotrifluorid
Formel:
C₇H₃ClF₄ Molecular Weight: 198.5453 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412198 CAS-Nummer: 103889-37-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...