tert-Butyl-2-(trifluoromethyl)acrylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4287-1G
PC4287-500MG
PC4287-5G
APOSPC4287-1GEA
95.7
EUR
APOSPC4287-1G
APOSPC4287-500MG
APOSPC4287-5G
tert-Butyl-2-(trifluoromethyl)acrylate 98%
tert-Butyl-2-(trifluoromethyl)acrylate
Formel:
C₈H₁₁F₃O₂ Molecular Weight: 196.17 g/mol Siedepunkt: 120 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04038275 CAS-Nummer: 105935-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...