Cer(III)trifluormethansulfonat 99%

Lieferant: Apollo Scientific
Achtung

PC4288-1G PC4288-5G
APOSPC4288-1GEA 21.9 EUR
APOSPC4288-1G APOSPC4288-5G
Cer(III)trifluormethansulfonat 99%
Cer(III)trifluormethansulfonat
Formel: C₃CeF₉O₉S₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02093899
CAS-Nummer: 76089-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR