4-Fluorthiophenol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Fluorbenzol-1-thiol, 4-Fluorphenylmercaptan, 1-Fluor-4-mercaptobenzol, 4-Mercaptofluorbenzol, 4-Fluorbenzolthiol, p-Fluorthiophenol
PC4300-250G
PC4300-100G
PC4300-25G
APOSPC4300-250GEA
87
EUR
APOSPC4300-250G
APOSPC4300-100G
APOSPC4300-25G
4-Fluorthiophenol 98%
4-Fluorthiophenol
Intermediate for the synthesis of Bicalutamide.
Formel:
C₆H₅FS Molecular Weight: 128,17 g/mol Siedepunkt: 168…172 °C Dichte: 1,203 g/cm³ (25 °C) Flammpunkt: 54 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004846 CAS-Nummer: 371-42-6 EINECS: 206-737-6 UN: 3071 ADR: 6.1+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...