4-Bromo-2-chloro-3-fluorophenol 95+%

Lieferant: Apollo Scientific

PC430005-250MG PC430005-1G
APOSPC430005-250MGEA 59.6 EUR
APOSPC430005-250MG APOSPC430005-1G
4-Bromo-2-chloro-3-fluorophenol 95+%
4-Bromo-2-chloro-3-fluorophenol
Formel: C₆H₃BrClFO
Molecular Weight: 225,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11849932
CAS-Nummer: 1233026-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR