tert-Butyl-3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate 95+%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate

PC430112-250MG PC430112-1G PC430112-5G
APOSPC430112-250MGEA 77 EUR
APOSPC430112-250MG APOSPC430112-1G APOSPC430112-5G
tert-Butyl-3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate 95+%
tert-Butyl-3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate
Formel: C₁₀H₁₆F₃NO₃
Molecular Weight: 255,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16990772
CAS-Nummer: 644970-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR