[6-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol

Lieferant: Apollo Scientific
Achtung

PC430532-1G PC430532-250MG
APOSPC430532-1GEA 102 EUR
APOSPC430532-1G APOSPC430532-250MG
[6-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol
[6-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol
Formel: C₁₀H₇F₃OS
Molecular Weight: 232,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11840966
CAS-Nummer: 863118-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR